Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 6 | 1925 | ||
GenBank mRNA | AK016482.1 | - | 6 | 1926 | ||
AK045422.1 | - | 6 | 1909 | |||
AK049394.1 | - | 6 | 1871 | |||
AK161312.1 | - | 6 | 1912 | |||
BC053515.1 | - | 6 | 1912 | |||
BC062139.1 | - | 6 | 1906 | 6334946 |