Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024499.1 | NP_078775.1 | 7 | 650 | ||
GenBank mRNA | AK039966.1 | - | 6 | 645 | ||
AK051005.1 | - | 7 | 649 | |||
AK079168.1 | - | 7 | 660 | |||
AK165557.1 | - | 7 | 674 | |||
BC003836.1 | - | 7 | 650 | 3486148 |