Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025305.2 | NP_079581.1 | 5 | 683 | ||
GenBank mRNA | AK009251.1 | - | 5 | 648 | ||
AK012225.1 | - | 5 | 683 | |||
AK131937.1 | - | 5 | 694 | |||
AK162024.1 | - | 5 | 662 | |||
BC049636.1 | - | 5 | 683 | 6532964 | ||
Y10968.1 | - | 4 | 568 |