Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025602.3 | NP_079878.1 | 3 | 569 | ||
GenBank mRNA | AK012917.1 | - | 3 | 552 | ||
AK019970.1 | - | 3 | 570 | |||
AK135550.1 | - | 3 | 594 | |||
BC027401.1 | - | 3 | 551 | 4948072 |