Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025918.3 | NP_080194.1 | 5 | 688 | ||
GenBank mRNA | AK009319.1 | - | 5 | 684 | ||
AK011254.1 | - | 5 | 689 | |||
AK028834.1 | - | 5 | 699 | |||
AK166820.1 | - | 5 | 684 | |||
BC061076.1 | - | 5 | 697 | 6594359 |