Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006802.3 | NP_006793.1 | 3, 4 | 345 | ||
GenBank mRNA | AK128438.1 | - | 3, 4 | 1154 | ||
AK222554.1 | - | 3, 4 | 221 | |||
AK291181.1 | - | 3, 4 | 226 | |||
AK292116.1 | - | 3, 4 | 256 | |||
BC002395.2 | - | 3, 4 | 238 | 2821350 | ||
BC011523.1 | - | 3, 4 | 302 | 3867442 | ||
DA835019.1 | - | 3, 4 | 345 | |||
U08815.1 | - | 3, 4 | 208 |