Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026187.5 | NP_080463.3 | 11 | 2216 | ||
GenBank mRNA | AK004948.2 | - | 9 | 2646 | ||
AK012672.1 | - | 4 | 789 | |||
AK156255.1 | - | 11 | 2178 | |||
AK157405.1 | - | 11 | 2178 | |||
AK194161.1 | - | 22 | ||||
BC039910.1 | - | 8 | 2866 | |||
BC057896.1 | - | 6 | 1058 | |||
BC071240.1 | - | 11 | 2146 | |||
BC150879.1 | - | 11 | 2166 | 9087790 |