Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI426949.1 | - | 1 | 99 |
Gene Symbol: Ccdc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026222.2 | NP_080498.1 | 16, 17 | 2391 | ||
GenBank mRNA | AK014839.1 | - | 16, 17 | 2383 | ||
AK030087.1 | - | 16, 17 | 2358 | |||
BC138917.1 | - | 16, 17 | 2353 | 8861939 | ||
BC138920.1 | - | 16, 17 | 2353 | 8861942 |