Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026792.3 | NP_081068.1 | 4, 5 | 772 | ||
GenBank mRNA | AK003649.1 | - | 4, 5 | 516 | ||
AK082137.1 | - | 4, 5 | 547 | |||
AK089885.1 | - | 4, 5 | 575 | |||
AK152709.1 | - | 4, 5 | 515 | |||
AK152889.1 | - | 4, 5 | 519 | |||
AK155378.1 | - | 5, 6 | 721 | |||
AK170093.1 | - | 4, 5 | 516 | |||
AY161042.1 | - | 4, 5 | 749 | |||
BC062903.1 | - | 4, 5 | 772 |