Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbndd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048227.1 | NP_001041692.1 | 4 | 677 | ||
NM_001048228.1 | NP_001041693.1 | 4 | 701 | |||
NM_001048229.1 | NP_001041694.1 | 3 | 704 | |||
NM_026797.2 | NP_081073.1 | 4 | 806 | |||
GenBank mRNA | AK003735.1 | - | 4 | 702 | ||
AK013571.1 | - | 4 | 612 | |||
AY187289.1 | - | 4 | 685 | |||
BC019947.1 | - | 4 | 708 | 4022474 | ||
BC030298.1 | - | 4 | 955 | 5364949 |