Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026819.3 | NP_081095.2 | 3 | 491 | ||
GenBank mRNA | AK003958.1 | - | 3 | 381 | ||
AK146317.1 | - | 3 | 438 | |||
AK159964.1 | - | 3 | 413 | |||
AK168115.1 | - | 3 | 438 | |||
AK168147.1 | - | 3 | 438 | |||
BC003930.1 | - | 3 | 422 | 3485584 | ||
BC023825.1 | - | 2 | 1533 | |||
BU963142.1 | - | 494 |