Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006810.3 | NP_006801.1 | 2, 3 | 335 | ||
NR_028444.1 | 2, 3 | 335 | ||||
GenBank mRNA | BC001625.1 | - | 2, 3 | 282 | 3537659 | |
BM549365.1 | - | 2, 3 | 335 | |||
D49490.1 | - | 2, 3 | 235 |