Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 6, 7 | 704 | ||
NM_028027.3 | NP_082303.2 | 6, 7 | 766 | |||
GenBank mRNA | AF487515.1 | - | 6, 7 | 732 | ||
AK010452.1 | - | 6, 7 | 732 | |||
AK158684.1 | - | 6, 7 | 704 | |||
AK210287.1 | - | 3, 4 | 261 | |||
BC041113.1 | - | 6, 7 | 748 | |||
CF724487.1 | - | 6, 7 | 651 |