Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028262.3 | NP_082538.2 | 4 | 475 | ||
GenBank mRNA | AK011993.1 | - | 4 | 455 | ||
AK031371.1 | - | 4 | 438 | |||
AK146777.1 | - | 4 | 469 | |||
AK166570.1 | - | 4 | 407 | |||
AY513271.1 | - | 4 | 408 | |||
BC016123.1 | - | 4 | 405 | 4503842 | ||
BC019973.1 | - | 4 | 405 | 4195130 | ||
BC108358.1 | - | 4 | 446 |