Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028731.5 | NP_083007.2 | 16 | 1832 | ||
GenBank mRNA | AK014813.1 | - | 18 | 1942 | ||
AK044360.1 | - | 16 | 1834 | |||
AK157352.1 | - | 16 | 1801 | |||
BC040392.1 | - | 6 | 515 | |||
BC052440.1 | - | 6 | 523 | |||
BC054797.1 | - | 3 | 485 | |||
BC059230.1 | - | 6 | 523 | |||
BC138937.1 | - | 16 | 1741 | 8861959 |