Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028771.2 | NP_083047.1 | 3 | 653 | ||
GenBank mRNA | AK004762.1 | - | 3 | 654 | ||
AK013304.1 | - | 3 | 604 | |||
AK045509.1 | - | 3 | 645 | |||
AK088221.1 | - | 3 | 644 | |||
AK155466.1 | - | 3 | 636 | |||
BC006752.1 | - | 2 | 158 | |||
BC078634.1 | - | 3 | 620 | 5388281 |