Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006816.2 | NP_006807.1 | 6 | 938 | ||
GenBank mRNA | AK222609.1 | - | 6 | 705 | ||
AK292099.1 | - | 6 | 715 | |||
AK301607.1 | - | 7 | 622 | |||
BC017263.2 | - | 6 | 718 | 4135210 | ||
U10362.1 | - | 6 | 701 |