Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033560.3 | NP_291038.2 | 3 | 617 | ||
GenBank mRNA | AK016385.1 | - | 3 | 599 | ||
AK048942.1 | - | 3 | 597 | |||
AK054055.1 | - | 3 | 599 | |||
AK137479.1 | - | 3 | 602 | |||
AK157507.1 | - | 3 | 583 | |||
AK170280.1 | - | 3 | 586 | |||
AK170943.1 | - | 3 | 575 | |||
AK170961.1 | - | 3 | 606 | |||
AK171055.1 | - | 3 | 606 | |||
AK171221.1 | - | 3 | 618 | |||
BC039290.1 | - | 3 | 573 | 5066508 | ||
BC110396.1 | - | 3 | 532 | 5253683 |