Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033560.3 | NP_291038.2 | 8 | 1215 | ||
GenBank mRNA | AK016385.1 | - | 8 | 1198 | ||
AK048942.1 | - | 7 | 1277 | |||
AK054055.1 | - | 8 | 1197 | |||
AK157507.1 | - | 8 | 1182 | |||
AK170280.1 | - | 8 | 1184 | |||
AK170943.1 | - | 8 | 1173 | |||
AK170961.1 | - | 8 | 1204 | |||
AK171055.1 | - | 8 | 1204 | |||
AK171221.1 | - | 8 | 1216 | |||
AK218075.1 | - | 3 | 216 | |||
BC039290.1 | - | 8 | 1171 | 5066508 | ||
BC110396.1 | - | 8 | 1130 | 5253683 |