Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053079.2 | NP_444309.2 | 17 | 1317 | ||
GenBank mRNA | AF205540.1 | - | 17 | 1316 | ||
AK085471.1 | - | 15 | 2424 | |||
AK136428.1 | - | 17 | 1318 | |||
BC116248.1 | - | 17 | 1307 | 40093541 | ||
BC116249.1 | - | 17 | 1307 | 40093542 |