Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 2, 3 | 686 | ||
XM_011540539.2 | XP_011538841.1 | 2, 3 | 524 | |||
GenBank mRNA | AB000115.1 | - | 2, 3 | 610 | ||
AK223087.1 | - | 2, 3 | 620 | |||
AK225442.1 | - | 2, 3 | 587 | |||
BC015932.2 | - | 2, 3 | 592 | 3909331 |