Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290430.1 | NP_001277359.1 | 8 | 783 | ||
NM_133991.2 | NP_598752.2 | 8 | 783 | |||
GenBank mRNA | AK036313.1 | - | 8 | 741 | ||
AK150251.1 | - | 6 | 759 | |||
BC011144.1 | - | 7 | 687 | 4194843 | ||
BC034880.1 | - | 8 | 653 | 3468633 |