Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134007.4 | NP_598768.1 | 3 | 376 | ||
GenBank mRNA | AK075907.1 | - | 3 | 377 | ||
AK131595.1 | - | 3 | 376 | |||
AK131605.1 | - | 3 | 355 | |||
AK131812.1 | - | 3 | 380 | |||
AK131995.1 | - | 3 | 387 | |||
AK167898.1 | - | 3 | 355 | |||
BC013522.1 | - | 3 | 365 | 4236502 | ||
BC019860.1 | - | 3 | 392 | 5123692 | ||
BC021952.1 | - | 3 | 413 | 5102315 |