Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700088E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138581.2 | NP_613047.1 | 2, 3 | 207 | ||
GenBank mRNA | AA008475.1 | - | 2, 3 | 196 | ||
AK006539.1 | - | 2, 3 | 188 | |||
AK007028.1 | - | 2, 3 | 214 | |||
BC068203.1 | - | 1, 2 | 252 | |||
BC096672.1 | - | 1, 2 | 305 | 5117646 | ||
BC116891.1 | - | 2, 3 | 138 | 40126210 | ||
BC116893.1 | - | 2, 3 | 138 | 40126212 | ||
BC144714.1 | - | 2, 3 | 138 | 9053262 |