Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138583.2 | NP_613049.2 | 6 | 510 | ||
GenBank mRNA | AK089239.1 | - | 6 | 492 | ||
AK133588.1 | - | 6 | 493 | |||
AK168595.1 | - | 5 | 489 | |||
AK199175.1 | - | 1 | 52 | |||
BC005445.1 | - | 6 | 571 | 3496749 | ||
X74504.1 | - | 5 | 483 |