Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006827.5 | NP_006818.3 | 3 | 468 | ||
GenBank mRNA | AJ004913.1 | - | 3 | 412 | ||
AK222972.1 | - | 3 | 420 | |||
AK302920.1 | - | 2 | 129 | |||
AK308864.1 | - | 1 | 87 | |||
AK310371.1 | - | 3 | 419 | |||
AK312384.1 | - | 3 | 437 | |||
AL832012.2 | - | 3 | 412 | |||
BC001496.2 | - | 3 | 453 | 2959718 | ||
BC001825.2 | - | 3 | 453 | 2959718 | ||
BX248754.1 | - | 3 | 419 | |||
BX504393.1 | - | 3 | 469 | |||
X97442.1 | - | 3 | 412 |