Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam104a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138598.5 | NP_613064.2 | 3 | 2112 | ||
GenBank mRNA | AK009763.2 | - | 3 | 1993 | ||
AK087540.1 | - | 3 | 2023 | |||
AK158872.1 | - | 3 | 2159 | |||
AK159228.1 | - | 3 | 2013 | |||
AK159659.1 | - | 3 | 2011 | |||
AK211621.1 | - | 1 | 23 | |||
BC025149.1 | - | 1 | 1125 | |||
BC056429.1 | - | 3 | 1854 |