Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138603.3 | NP_613069.3 | 10 | 1235 | ||
GenBank mRNA | AF229637.1 | - | 10 | 1176 | ||
AK010536.1 | - | 10 | 1153 | |||
AK039573.1 | - | 10 | 1208 | |||
BC011195.1 | - | 10 | 1140 | 4188459 | ||
BC057686.1 | - | 1 | 1285 |