Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144833.3 | NP_659082.1 | 11, 12 | 1866 | ||
GenBank mRNA | AF295806.1 | - | 11, 12 | 1628 | ||
AK051939.1 | - | 9, 10 | 1266 | |||
AK083003.1 | - | 11, 12 | 1609 | |||
AK132345.1 | - | 11, 12 | 1605 | |||
BC021528.1 | - | 11, 12 | 1621 | 5353673 | ||
BC056957.1 | - | 11, 12 | 1563 |