Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144884.2 | NP_659133.1 | 3 | 627 | ||
GenBank mRNA | AJ298841.1 | - | 3 | 582 | ||
AK038891.1 | - | 3 | 585 | |||
AK079728.1 | - | 2 | 899 | |||
AK082936.1 | - | 3 | 537 | |||
AK160361.1 | - | 3 | 583 | |||
BC017683.1 | - | 3 | 607 | 4238480 |