Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 7 | 874 | ||
GenBank mRNA | AK011822.1 | - | 7 | 861 | ||
AK042644.1 | - | 6 | 546 | |||
AK149957.1 | - | 7 | 707 | |||
AK177661.1 | - | 2 | 182 | |||
BC011161.1 | - | 7 | 835 | |||
BC019384.1 | - | 7 | 855 | 5123736 | ||
BC100575.1 | - | 7 | 806 | 6432108 |