Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170341.1 | NP_001163812.1 | 9 | 1181 | ||
NM_145385.2 | NP_663360.1 | 9 | 1202 | |||
GenBank mRNA | AK141241.1 | - | 9 | 1137 | ||
AK143972.1 | - | 9 | 1138 | |||
AK171220.1 | - | 2059 | ||||
AK194002.1 | - | 1 | 47 | |||
AK195037.1 | - | 1 | 49 | |||
AK207591.1 | - | 1 | 42 | |||
AK209928.1 | - | 1 | 42 | |||
BC003975.1 | - | 9 | 1113 | 3488851 | ||
CD560861.1 | - | 2 | 316 |