Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145528.3 | NP_663503.1 | 10 | 1111 | ||
GenBank mRNA | AK122339.1 | - | 6 | 830 | ||
AK149672.1 | - | 10 | 1070 | |||
AK154470.1 | - | 10 | 1099 | |||
AK163962.1 | - | 10 | 1065 | |||
AK171206.1 | - | 10 | 1097 | |||
AK171440.1 | - | 10 | 1088 | |||
BC016669.1 | - | 10 | 1095 | 2648797 | ||
BC023673.1 | - | 10 | 1061 | 5343236 | ||
BC023712.1 | - | 10 | 1084 | 5346303 | ||
BC033419.1 | - | 10 | 1062 | 5359944 |