Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145928.1 | NP_666040.1 | 9 | 1159 | ||
GenBank mRNA | AK030593.1 | - | 9 | 1161 | ||
AK156671.1 | - | 9 | 1172 | |||
BC024611.1 | - | 9 | 1164 | 4188261 | ||
BC025568.1 | - | 9 | 1173 | 5322531 | ||
BC026574.1 | - | 9 | 1159 | 4954874 |