Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 4 | 792 | ||
NM_001100170.1 | NP_001093640.1 | 4 | 792 | |||
NM_006839.2 | NP_006830.2 | 4 | 792 | |||
XM_005264110.3 | XP_005264167.1 | 4 | 464 | |||
XM_005264113.1 | XP_005264170.1 | 4 | 460 | |||
XM_005264114.1 | XP_005264171.1 | 4 | 464 | |||
XM_011532497.2 | XP_011530799.1 | 4 | 464 | |||
XM_017003194.1 | XP_016858683.1 | 4 | 460 | |||
XM_017003195.1 | XP_016858684.1 | 4 | 460 | |||
GenBank mRNA | AK225780.1 | - | 4 | 459 | ||
AK296676.1 | - | 4 | 434 | |||
AK299014.1 | - | 4 | 455 | |||
AK299596.1 | - | 2 | 202 | |||
AK300017.1 | - | 4 | 432 | |||
AK304197.1 | - | 4 | 469 | |||
AK312251.1 | - | 4 | 442 | |||
AY232292.1 | - | 4 | 425 | |||
AY921637.1 | - | 4 | 440 | |||
BC002412.2 | - | 4 | 433 | 2964676 | ||
BC005983.1 | - | 4 | 484 | |||
BC107419.1 | - | 4 | 488 | |||
BX537369.1 | - | 4 | 495 | |||
CR456793.1 | - | 4 | 404 | |||
D21092.1 | - | 2 | 294 | |||
D21094.1 | - | 4 | 489 | |||
L42572.1 | - | 4 | 496 |