Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173036.3 | NP_766624.3 | 12 | 2129 | ||
GenBank mRNA | AK014445.1 | - | 2 | 785 | ||
AK154648.1 | - | 12 | 2105 | |||
BC038936.1 | - | 1 | 571 | |||
BC050004.1 | - | 2 | 679 | |||
BC064726.1 | - | 12 | 2089 | 30433392 |
Set Membership: |
G-protein coupled receptor |