Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290624.1 | NP_001277553.1 | 5 | 606 | ||
GenBank mRNA | AK037217.1 | - | 5 | 569 | ||
AK083704.1 | - | 4 | 573 | |||
BC023671.1 | - | 5 | 602 | |||
BC032173.1 | - | 2 | 927 | |||
BC049258.1 | - | 5 | 548 | 5250488 |