Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297655.1 | NP_001284584.1 | 2, 3 | 279 | ||
NM_006845.3 | NP_006836.2 | 2, 3 | 279 | |||
XM_011540540.2 | XP_011538842.1 | 2, 3 | 393 | |||
GenBank mRNA | AB264115.1 | - | 2, 3 | 279 | ||
AK290688.1 | - | 2, 3 | 205 | |||
AK300756.1 | - | 2, 3 | 236 | |||
AK312606.1 | - | 2, 3 | 235 | |||
AU130795.1 | - | 2, 3 | 279 | |||
BC008764.2 | - | 2, 3 | 218 | 3347571 | ||
BC014924.1 | - | 2, 3 | 216 | 3909438 | ||
BT006759.1 | - | 2, 3 | 164 | |||
CR450302.1 | - | 2, 3 | 164 | |||
U63743.1 | - | 2, 3 | 218 |