Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177472.5 | NP_803423.2 | 3, 4 | 724 | ||
GenBank mRNA | AK173263.1 | - | 1, 2 | 19 | ||
BC065125.1 | - | 3, 4 | 729 | 30534878 |