Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D19Bwg1357e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031451.1 | - | 2 | 252 | ||
AK050829.1 | - | 9 | 956 | |||
AK088081.1 | - | 9 | 951 | |||
AK141893.1 | - | 9 | 954 | |||
AK172872.1 | - | 5 | 462 | |||
BC016186.1 | - | 9 | 969 | |||
BC041111.1 | - | 8 | 934 | |||
BC052686.1 | - | 8 | 907 | |||
BC145533.1 | - | 9 | 949 |