Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Imp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178601.3 | NP_848716.2 | 5 | 464 | ||
GenBank mRNA | AF334609.1 | - | 4 | 368 | ||
AK004135.1 | - | 4 | 366 | |||
AK008178.1 | - | 5 | 426 | |||
AK010504.1 | - | 5 | 424 | |||
AK013334.1 | - | 5 | 393 | |||
AK018039.1 | - | 5 | 422 | |||
AK084933.1 | - | 5 | 400 | |||
AK151176.1 | - | 5 | 443 | |||
AK152214.1 | - | 5 | 443 | |||
AK166903.1 | - | 5 | 414 | |||
BC037602.1 | - | 4 | 385 | |||
BC069930.1 | - | 4 | 374 | 3481801 | ||
DV649342.1 | - | 5 | 464 |