Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178607.4 | NP_848722.1 | 6 | 919 | ||
GenBank mRNA | AK015699.1 | - | 7 | 979 | ||
AK048813.1 | - | 6 | 877 | |||
AK083665.1 | - | 6 | 909 | |||
AK089207.1 | - | 6 | 906 | |||
AK154093.1 | - | 6 | 856 | |||
AK165427.1 | - | 6 | 877 | |||
BC096529.1 | - | 6 | 852 | 6406985 |