Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lair1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113474.1 | NP_001106945.1 | 4, 5 | 626 | ||
NM_178611.6 | NP_848726.1 | 3, 4 | 296 | |||
GenBank mRNA | AF479685.1 | - | 4, 5 | 561 | ||
AF491829.1 | - | 4, 5 | 659 | |||
AF491830.1 | - | 3, 4 | 208 | |||
AF491831.1 | - | 4, 5 | 538 | |||
AK033472.1 | - | 3, 4 | 273 | |||
AK038574.1 | - | 3, 4 | 259 | |||
AK043664.1 | - | 3, 4 | 353 | |||
AK044465.1 | - | 4, 5 | 597 | |||
AK050281.1 | - | 4, 5 | 582 | |||
AK054158.1 | - | 4, 5 | 705 | |||
AY392763.1 | - | 4, 5 | 563 | |||
AY392764.1 | - | 3, 4 | 233 | |||
BC025901.1 | - | 4, 5 | 590 | 5253846 | ||
BY204263.1 | - | 2, 3 | 240 |