Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006717.2 | NP_006708.2 | 6 | 1348 | ||
GenBank mRNA | AF087864.1 | - | 6 | 1228 | ||
AF106682.1 | - | 6 | 1229 | |||
AF317228.2 | - | 5 | 1193 | |||
AK092017.1 | - | 9 | 1690 | |||
AK290009.1 | - | 6 | 1216 | |||
AL136719.1 | - | 6 | 1311 | |||
BC013571.2 | - | 6 | 1332 |