Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254750.1 | NP_001241679.1 | 9, 10 | 1719 | ||
NM_001254751.1 | NP_001241680.1 | 9, 10 | 1692 | |||
NM_006725.4 | NP_006716.3 | 10, 11 | 1815 | |||
NR_045638.1 | 10, 11 | 1795 | ||||
XM_006718738.1 | XP_006718801.1 | 9, 10 | 1719 | |||
XM_006718739.1 | XP_006718802.1 | 9, 10 | 1692 | |||
XM_006718740.1 | XP_006718803.1 | 9, 10 | 1692 | |||
XM_006718741.1 | XP_006718804.1 | 8, 9 | 1596 | |||
XM_011545360.1 | XP_011543662.1 | 10, 11 | 1815 | |||
XM_011545362.1 | XP_011543664.1 | 9, 10 | 1512 | |||
GenBank mRNA | AK128419.1 | - | 11, 12 | 1719 | ||
AK292704.1 | - | 10, 11 | 1691 | |||
AK312749.1 | - | 10, 11 | 1776 | |||
BC033755.1 | - | 10, 11 | 1731 | 5213263 | ||
BC078669.1 | - | 10, 11 | 1754 | 6302947 | ||
U66142.1 | - | 10, 11 | 1590 | |||
U66145.1 | - | 9, 10 | 1494 | |||
U66146.1 | - | 9, 10 | 1467 | |||
X60992.1 | - | 10, 11 | 1690 |