Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198937.2 | NP_945175.1 | 5 | 2293 | ||
GenBank mRNA | AK150226.1 | - | 5 | 2305 | ||
AK152133.1 | - | 5 | 2309 | |||
AK152573.1 | - | 5 | 2307 | |||
AK166213.1 | - | 5 | 2303 | |||
AK167223.1 | - | 5 | 2277 | |||
BC057024.1 | - | 5 | 2291 | 6815840 | ||
BC080740.1 | - | 6 | 2221 | 30460749 |