Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163018.1 | NP_001156490.1 | 6 | 884 | ||
NM_199196.2 | NP_954666.1 | 7 | 953 | |||
GenBank mRNA | AK122213.1 | - | 7 | 938 | ||
BC039915.1 | - | 1 | 29 | |||
BC064461.1 | - | 7 | 953 | 6821922 | ||
BC084591.1 | - | 2 | 224 |