Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007593.5 | NP_031619.3 | 1 | 819 | ||
GenBank mRNA | AK005837.1 | - | 1 | 820 | ||
AK006940.1 | - | 1 | 820 | |||
AK015808.1 | - | 818 | ||||
AK076986.1 | - | 829 |