Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009313.5 | NP_033339.2 | 6 | 2263 | ||
GenBank mRNA | AK038558.1 | - | 5 | 1920 | ||
BC075631.1 | - | 6 | 2263 | 30536972 | ||
BC098332.1 | - | 5 | 2170 | 30618488 |
Set Membership: |
G-protein coupled receptor |