Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009315.3 | NP_033341.1 | 10 | 994 | ||
GenBank mRNA | AK153731.1 | - | 1042 | |||
AK154867.1 | - | 10 | 1028 | |||
BC058583.1 | - | 10 | 994 | 6514960 | ||
D49439.1 | - | 9 | 927 |